Product Details

SNP ID
rs143394852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76035314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTACCCCAGGGCCTATGCAAAG[A/G]CAGCGTCCTCTTTTGAACATGGGGA
Phenotype
MIM: 603086 MIM: 604852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ART3 PubMed Links

Gene Details

Gene
ART3
Gene Name
ADP-ribosyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130016.2 300 Intron NP_001123488.1
NM_001130017.2 300 Intron NP_001123489.1
NM_001179.5 300 Intron NP_001170.2
XM_005262997.1 300 Intron XP_005263054.1
XM_005262999.1 300 Intron XP_005263056.1
XM_005263003.1 300 Intron XP_005263060.1
XM_005263004.1 300 Intron XP_005263061.1
XM_006714220.1 300 Intron XP_006714283.1
XM_011531971.1 300 Intron XP_011530273.1
XM_017008206.1 300 Intron XP_016863695.1
XM_017008207.1 300 Intron XP_016863696.1
XM_017008208.1 300 Intron XP_016863697.1
XM_017008209.1 300 Intron XP_016863698.1
XM_017008210.1 300 Intron XP_016863699.1
Gene
CXCL11
Gene Name
C-X-C motif chemokine ligand 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302123.1 300 Silent Mutation TGC,TGT C30C NP_001289052.1
NM_005409.4 300 Silent Mutation TGC,TGT C30C NP_005400.1

View Full Product Details