Product Details

SNP ID
rs143489137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:20728684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCACATTGAGTTTACACAAACA[C/T]GTGATGGCAAATTTCAGTGTACATG
Phenotype
MIM: 608182
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNIP4 PubMed Links

Gene Details

Gene
KCNIP4
Gene Name
potassium voltage-gated channel interacting protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035003.1 2184 UTR 3 NP_001030175.1
NM_001035004.1 2184 UTR 3 NP_001030176.1
NM_025221.5 2184 UTR 3 NP_079497.2
NM_147181.3 2184 UTR 3 NP_671710.1
NM_147182.3 2184 UTR 3 NP_671711.1
NM_147183.3 2184 UTR 3 NP_671712.1
XM_005248190.4 2184 UTR 3 XP_005248247.1
XM_011513882.2 2184 UTR 3 XP_011512184.1
XM_011513883.2 2184 UTR 3 XP_011512185.1
XM_011513884.2 2184 UTR 3 XP_011512186.1
XM_011513885.2 2184 UTR 3 XP_011512187.1
XM_011513886.2 2184 UTR 3 XP_011512188.1
XM_011513887.2 2184 UTR 3 XP_011512189.1
XM_011513889.2 2184 UTR 3 XP_011512191.1
XM_017008653.1 2184 UTR 3 XP_016864142.1
Gene
PACRGL
Gene Name
PARK2 coregulated like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130727.1 2184 Intron NP_001124199.1
NM_001258345.1 2184 Intron NP_001245274.1
NM_001258346.1 2184 Intron NP_001245275.1
NM_001317849.1 2184 Intron NP_001304778.1
NM_145048.3 2184 Intron NP_659485.1
XM_011513780.1 2184 Intron XP_011512082.1
XM_011513781.1 2184 Intron XP_011512083.1
XM_011513784.1 2184 Intron XP_011512086.1
XM_011513785.1 2184 Intron XP_011512087.1
XM_011513786.2 2184 UTR 3 XP_011512088.1
XM_011513787.1 2184 Intron XP_011512089.1
XM_011513788.1 2184 Intron XP_011512090.1
XM_011513791.1 2184 Intron XP_011512093.1
XM_011513792.1 2184 Intron XP_011512094.1
XM_011513794.1 2184 Intron XP_011512096.1
XM_011513795.1 2184 Intron XP_011512097.1
XM_011513796.1 2184 Intron XP_011512098.1
XM_011513797.2 2184 Intron XP_011512099.1
XM_011513798.2 2184 Intron XP_011512100.1
XM_011513804.1 2184 Intron XP_011512106.1
XM_011513805.2 2184 UTR 3 XP_011512107.1
XM_017007747.1 2184 UTR 3 XP_016863236.1
XM_017007748.1 2184 UTR 3 XP_016863237.1
XM_017007749.1 2184 UTR 3 XP_016863238.1
XM_017007750.1 2184 Intron XP_016863239.1
XM_017007751.1 2184 UTR 3 XP_016863240.1
XM_017007752.1 2184 UTR 3 XP_016863241.1
XM_017007753.1 2184 UTR 3 XP_016863242.1
XM_017007754.1 2184 UTR 3 XP_016863243.1
XM_017007755.1 2184 UTR 3 XP_016863244.1
XM_017007756.1 2184 UTR 3 XP_016863245.1
XM_017007757.1 2184 UTR 3 XP_016863246.1
XM_017007758.1 2184 UTR 3 XP_016863247.1
XM_017007759.1 2184 UTR 3 XP_016863248.1

View Full Product Details