Product Details
- SNP ID
-
rs143568587
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:166735071 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAATCGCCACTAGCAAAATCTCC[A/G]GAGATCTCAAAATCTATAGCTTAAA
- Phenotype
-
MIM: 607989
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPOCK3
PubMed Links
Gene Details
- Gene
- SPOCK3
- Gene Name
- sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040159.1 |
1188 |
Silent Mutation |
TCC,TCT |
S384S |
NP_001035249.1 |
NM_001204352.1 |
1188 |
Silent Mutation |
TCC,TCT |
S336S |
NP_001191281.1 |
NM_001204353.1 |
1188 |
Silent Mutation |
TCC,TCT |
S267S |
NP_001191282.1 |
NM_001204354.1 |
1188 |
Silent Mutation |
TCC,TCT |
S295S |
NP_001191283.1 |
NM_001204355.1 |
1188 |
Silent Mutation |
TCC,TCT |
S291S |
NP_001191284.1 |
NM_001204356.1 |
1188 |
Silent Mutation |
TCC,TCT |
S344S |
NP_001191285.1 |
NM_001204358.1 |
1188 |
Intron |
|
|
NP_001191287.1 |
NM_001251967.1 |
1188 |
Silent Mutation |
TCC,TCT |
S289S |
NP_001238896.1 |
NM_016950.2 |
1188 |
Silent Mutation |
TCC,TCT |
S387S |
NP_058646.2 |
XM_011532018.1 |
1188 |
Silent Mutation |
TCC,TCT |
S387S |
XP_011530320.1 |
XM_017008257.1 |
1188 |
Silent Mutation |
TCC,TCT |
S396S |
XP_016863746.1 |
XM_017008258.1 |
1188 |
Silent Mutation |
TCC,TCT |
S384S |
XP_016863747.1 |
View Full Product Details