Product Details

SNP ID
rs143604530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140561725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTACACAACACTTGTTTCTAG[A/G]TGAGTGTAATAATTACCCAAATCAA
Phenotype
MIM: 610196 MIM: 113730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELMOD2 PubMed Links

Gene Details

Gene
ELMOD2
Gene Name
ELMO domain containing 2
There are no transcripts associated with this gene.

Gene
UCP1
Gene Name
uncoupling protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021833.4 Intron NP_068605.1
XM_005263206.3 Intron XP_005263263.1
XM_011532228.2 Intron XP_011530530.1

View Full Product Details