Product Details

SNP ID
rs143794658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155831831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTAAATAACACTTACCAAGTAA[C/G]TCAGACACACTCACCGCCTTTTGCT
Phenotype
MIM: 616693
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASIC5 PubMed Links

Gene Details

Gene
ASIC5
Gene Name
acid sensing ion channel subunit family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017419.2 1367 Missense Mutation GAC,GAG D440E NP_059115.1
XM_017008291.1 1367 Missense Mutation GAC,GAG D398E XP_016863780.1

View Full Product Details