Product Details

SNP ID
rs143817654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188139762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGCCCGCATTTCCAGTCAAA[C/T]GAGCGTCTGGGGAGGCTGGCCAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIML1 PubMed Links

Gene Details

Gene
TRIML1
Gene Name
tripartite motif family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178556.3 319 Silent Mutation AAC,AAT N68N NP_848651.2
XM_006714207.3 319 Silent Mutation AAC,AAT N68N XP_006714270.1
XM_011531919.1 319 UTR 5 XP_011530221.1

View Full Product Details