Product Details

SNP ID
rs143875438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121036593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACGGTGGCTGAGGAACAGGTAC[A/G]GAGCTTGTCAAAGGCTTTGATTCTT
Phenotype
MIM: 616506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDNF PubMed Links

Gene Details

Gene
NDNF
Gene Name
neuron derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024574.3 1905 Missense Mutation CGT,TGT R460C NP_078850.3

View Full Product Details