Product Details

SNP ID
rs143978776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70335493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTCTTAGCTACCATTGACACA[A/G]GAGATGCAGAGATCTCAATAACCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABS1 PubMed Links

Gene Details

Gene
CABS1
Gene Name
calcium binding protein, spermatid associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033122.3 540 Missense Mutation AGA,GGA R152G NP_149113.3

View Full Product Details