Product Details

SNP ID
rs144030490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:785026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACGCAGGGGCCCCGACATGCAG[A/G]AGGAGGCGCAGGCGCAGGACAGACG
Phenotype
MIM: 605032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPLX1 PubMed Links

Gene Details

Gene
CPLX1
Gene Name
complexin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006651.3 2002 UTR 3 NP_006642.1
XM_011513391.1 2002 UTR 3 XP_011511693.1
Gene
LOC100129917
Gene Name
uncharacterized LOC100129917
There are no transcripts associated with this gene.

View Full Product Details