Product Details

SNP ID
rs144040737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152770003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAGAGCTTAAAAATTTCTTGAT[A/G]AGACAGTGTCGATATTTGATTTTAA
Phenotype
MIM: 605928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFIP1 PubMed Links

Gene Details

Gene
ARFIP1
Gene Name
ADP ribosylation factor interacting protein 1
There are no transcripts associated with this gene.

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 1823 Silent Mutation CTC,CTT L334L NP_663772.1
XM_005262807.4 1823 Silent Mutation CTC,CTT L334L XP_005262864.1

View Full Product Details