Product Details

SNP ID
rs144132835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68228006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTACCACATCCATCACCCCAGC[G/T]TACTATTCCAACAAGATGCCAGATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11B PubMed Links

Gene Details

Gene
TMPRSS11B
Gene Name
transmembrane protease, serine 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182502.3 1292 Missense Mutation AGC,CGC S386R NP_872308.2
XM_011531608.2 1292 Missense Mutation AGC,CGC S386R XP_011529910.1
XM_011531609.1 1292 Intron XP_011529911.1
Gene
TMPRSS11BNL
Gene Name
TMPRSS11B N-terminal like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details