Product Details
- SNP ID
-
rs144214818
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:77048398 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTACTTTGCGTTTAGTAGAGG[A/T]CTGCTTGCACCCACTGGCAGCTGGG
- Phenotype
-
MIM: 612887
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCNI
PubMed Links
Gene Details
- Gene
- CCNI
- Gene Name
- cyclin I
- Gene
- SEPT11
- Gene Name
- septin 11
There are no transcripts associated with this gene.
View Full Product Details