Product Details
- SNP ID
-
rs144507683
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:183100562 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCAGGGATAACTTCGACATTTGG[A/G]CTTTTGTCATTTCTGAAAATTATAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WWC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146320791] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WWC2
- Gene Name
- WW and C2 domain containing 2
- Gene
- WWC2-AS2
- Gene Name
- WWC2 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details