Product Details

SNP ID
rs144624268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5904748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTAGGAAAGAGTTCACATGCA[C/T]ACAACTTTTTGTGCAGGTTGGGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4orf50 PubMed Links

Gene Details

Gene
C4orf50
Gene Name
chromosome 4 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011513603.2 7317 UTR 3 XP_011511905.1
XM_011513604.2 7317 UTR 3 XP_011511906.1
XM_017008891.1 7317 UTR 3 XP_016864380.1
XM_017008892.1 7317 UTR 3 XP_016864381.1
XM_017008893.1 7317 UTR 3 XP_016864382.1
XM_017008894.1 7317 Intron XP_016864383.1

View Full Product Details