Product Details

SNP ID
rs144675086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158715616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATCTGGATGACTGTCGCCAGA[C/G]CCATCTTTTGGGAATATTCCCCCGT
Phenotype
MIM: 231675 MIM: 601753
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ETFDH PubMed Links

Gene Details

Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
There are no transcripts associated with this gene.

Gene
PPID
Gene Name
peptidylprolyl isomerase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005038.2 703 Silent Mutation GGC,GGG G197G NP_005029.1

View Full Product Details