Product Details

SNP ID
rs145176475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70819663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGGTATCACCTGTCAGAAACC[C/T]GGGCAAATTTAATTTTAAGAACTAG
Phenotype
MIM: 604851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRSF1 PubMed Links

Gene Details

Gene
GRSF1
Gene Name
G-rich RNA sequence binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098477.1 2586 UTR 3 NP_001091947.1
NM_002092.3 2586 UTR 3 NP_002083.3
XM_005265681.3 2586 UTR 3 XP_005265738.1
XM_011531897.2 2586 UTR 3 XP_011530199.1

View Full Product Details