Product Details

SNP ID
rs145194479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:114609876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTTGATTTCCTGGTGGCCTGT[A/G]CAGTGAGTATTTTGTAACATTTAGA
Phenotype
MIM: 601291
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT8 PubMed Links

Gene Details

Gene
UGT8
Gene Name
UDP glycosyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128174.2 Intron NP_001121646.1
NM_001322112.1 Intron NP_001309041.1
NM_001322113.1 Intron NP_001309042.1
NM_001322114.1 Intron NP_001309043.1
NM_003360.4 Intron NP_003351.2

View Full Product Details