Product Details

SNP ID
rs145208075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1395645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTGCCCGCCTGCTCACACGTG[A/C]CCATGTGGAGTGCCTGCCTGCTCAC
Phenotype
MIM: 610203 MIM: 614632
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRIPAK PubMed Links

Gene Details

Gene
CRIPAK
Gene Name
cysteine rich PAK1 inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175918.3 4094 Nonsense Mutation TGA,TGC *378C NP_787114.2
Gene
NKX1-1
Gene Name
NK1 homeobox 1
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
There are no transcripts associated with this gene.

View Full Product Details