Product Details

SNP ID
rs145215771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154744860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCACCCCGATCAGTCCCCAGTC[C/T]GACAAGGTATGATGTGTGACTCCCA
Phenotype
MIM: 604863
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRAT PubMed Links

Gene Details

Gene
LRAT
Gene Name
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301645.1 593 Silent Mutation TCC,TCT S178S NP_001288574.1
NM_004744.4 593 Silent Mutation TCC,TCT S178S NP_004735.2

View Full Product Details