Product Details

SNP ID
rs145220003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118723062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGCTATGCAGACCTTAAGTTGA[A/C]AAATGTGTTCAATGGAGTTACATGG
Phenotype
MIM: 607186
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC24D PubMed Links

Gene Details

Gene
SEC24D
Gene Name
SEC24 homolog D, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318066.1 3007 UTR 3 NP_001304995.1
NM_014822.3 3007 UTR 3 NP_055637.2
XM_005263379.2 3007 UTR 3 XP_005263436.1
XM_017008875.1 3007 UTR 3 XP_016864364.1

View Full Product Details