Product Details

SNP ID
rs145611966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186191922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGCCAGTCTGGTCCTGAGCCT[A/G]CTGCAGAGGGTGGCGAGCTACGCGC
Phenotype
MIM: 608614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP4V2 PubMed Links

Gene Details

Gene
CYP4V2
Gene Name
cytochrome P450 family 4 subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207352.3 188 Silent Mutation CTA,CTG L33L NP_997235.3
XM_005262935.3 188 Silent Mutation CTA,CTG L33L XP_005262992.1
XM_017008037.1 188 UTR 5 XP_016863526.1
Gene
FLJ38576
Gene Name
uncharacterized LOC651430
There are no transcripts associated with this gene.

View Full Product Details