Product Details

SNP ID
rs145737909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47453180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATGCAGGCTTAAAAGTGGCATT[C/T]GTAATGCAGCAATCTTGTCACTGGA
Phenotype
MIM: 616656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COMMD8 PubMed Links

Gene Details

Gene
COMMD8
Gene Name
COMM domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017008330.1 461 Missense Mutation CAA,CGA Q137R XP_016863819.1

View Full Product Details