Product Details

SNP ID
rs145802010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174644042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCCATAGGCTGTGAAGCTGAAT[C/T]GGCTTTCCCTTACCTCATCATCCTG
Phenotype
MIM: 600421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRA3 PubMed Links

Gene Details

Gene
GLRA3
Gene Name
glycine receptor alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042543.2 1643 Missense Mutation CAA,CGA Q365R NP_001036008.1
NM_006529.3 1643 Missense Mutation CAA,CGA Q380R NP_006520.2
XM_011532267.2 1643 Intron XP_011530569.1
XM_011532268.2 1643 Missense Mutation CAA,CGA Q185R XP_011530570.1
XM_017008630.1 1643 Missense Mutation CAA,CGA Q340R XP_016864119.1

View Full Product Details