Product Details

SNP ID
rs145869884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99905664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTAATAATGACACGAGGATCA[A/G]TACAATTATGGGCATCAGCTGGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 1314 Silent Mutation CTG,TTG L259L NP_001026893.1
NM_001278310.1 1314 Silent Mutation CTG,TTG L192L NP_001265239.1
NM_001278311.1 1314 Intron NP_001265240.1
XM_011532262.2 1314 Silent Mutation CTG,TTG L174L XP_011530564.1
XM_011532263.2 1314 Silent Mutation CTG,TTG L174L XP_011530565.1
XM_011532265.2 1314 Silent Mutation CTG,TTG L171L XP_011530567.1
XM_017008628.1 1314 Silent Mutation CTG,TTG L171L XP_016864117.1

View Full Product Details