Product Details

SNP ID
rs145901519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76309307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGACACAGAAGCTCCAGCTAC[C/T]TCTGAGACCAGTAACTCTAGGAGTT
Phenotype
MIM: 607406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC158 PubMed Links

Gene Details

Gene
CCDC158
Gene Name
coiled-coil domain containing 158
There are no transcripts associated with this gene.

Gene
FAM47E-STBD1
Gene Name
FAM47E-STBD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242939.1 1128 UTR 3 NP_001229868.1
Gene
STBD1
Gene Name
starch binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003943.4 1128 Silent Mutation ACC,ACT T128T NP_003934.1

View Full Product Details