Product Details

SNP ID
rs146019934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67821724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTTGCCTTAAATAGCTGAAAA[G/T]TATCCACTATTTCCTATGGAAGCAT
Phenotype
MIM: 605369
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11D PubMed Links

Gene Details

Gene
TMPRSS11D
Gene Name
transmembrane protease, serine 11D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004262.2 1935 UTR 3 NP_004253.1
XM_005265710.4 1935 UTR 3 XP_005265767.1
XM_017008851.1 1935 UTR 3 XP_016864340.1

View Full Product Details