Product Details

SNP ID
rs146200909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37590833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATCTGCTTTAATGAGAAGGGT[C/G]CTGTTCATGCCATGCCTGTGGTTGA
Phenotype
MIM: 611212
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C4orf19 PubMed Links

Gene Details

Gene
C4orf19
Gene Name
chromosome 4 open reading frame 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104629.1 1025 Missense Mutation CCT,GCT P260A NP_001098099.1
NM_018302.2 1025 Missense Mutation CCT,GCT P260A NP_060772.2
XM_011513712.2 1025 Missense Mutation CCT,GCT P260A XP_011512014.1
XM_011513713.2 1025 Missense Mutation CCT,GCT P260A XP_011512015.1
XM_017008361.1 1025 Missense Mutation CCT,GCT P260A XP_016863850.1
Gene
RELL1
Gene Name
RELT like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085399.1 1025 UTR 3 NP_001078868.1
NM_001085400.1 1025 Intron NP_001078869.1
XM_017008590.1 1025 Intron XP_016864079.1

View Full Product Details