Product Details
- SNP ID
-
rs146406150
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70053100 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATGAAGTTTTTTGTCTTTGCTTT[A/C]GTCTTGGCTCTCATGATTTCCATGA
- Phenotype
-
MIM: 142701
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HTN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73823840] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTN1
- Gene Name
- histatin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002159.3 |
118 |
Missense Mutation |
TTA,TTC |
L8F |
NP_002150.1 |
View Full Product Details