Product Details

SNP ID
rs146502138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82906377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAAGACAAAAAATCCTAAAACA[A/G]CCTCTTCCAGACAATTCTCAAGAAG
Phenotype
MIM: 610257 MIM: 612537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC31A PubMed Links

Gene Details

Gene
SEC31A
Gene Name
SEC31 homolog A, COPII coat complex component
There are no transcripts associated with this gene.

Gene
SNORD144
Gene Name
small nucleolar RNA, C/D box 144
There are no transcripts associated with this gene.

Gene
THAP9
Gene Name
THAP domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317776.1 430 UTR 5 NP_001304705.1
NM_024672.5 430 Silent Mutation CAA,CAG Q110Q NP_078948.3
XM_005263220.2 430 UTR 5 XP_005263277.1
XM_011532246.2 430 UTR 5 XP_011530548.1
XM_017008603.1 430 Silent Mutation CAA,CAG Q123Q XP_016864092.1
XM_017008604.1 430 UTR 5 XP_016864093.1
XM_017008605.1 430 UTR 5 XP_016864094.1
XM_017008606.1 430 UTR 5 XP_016864095.1
XM_017008607.1 430 UTR 5 XP_016864096.1
XM_017008608.1 430 UTR 5 XP_016864097.1
XM_017008609.1 430 UTR 5 XP_016864098.1
XM_017008610.1 430 UTR 5 XP_016864099.1
Gene
THAP9-AS1
Gene Name
THAP9 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details