Product Details

SNP ID
rs146571992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48488609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTTCCATCTTCCACCCAACTG[C/T]AAGAGGACTGTATGTCTGGAAACAG
Phenotype
MIM: 607520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
There are no transcripts associated with this gene.

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152679.3 1203 Silent Mutation TGC,TGT C328C NP_689892.1
Gene
ZAR1
Gene Name
zygote arrest 1
There are no transcripts associated with this gene.

View Full Product Details