Product Details

SNP ID
rs146973043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17843316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTATTTAGTGATCATGAAGTTC[C/G]AGAACCAGAATCAGAAATGAAGATG
Phenotype
MIM: 611799 MIM: 606280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LCORL PubMed Links

Gene Details

Gene
LCORL
Gene Name
ligand dependent nuclear receptor corepressor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166139.2 8217 Intron NP_001159611.1
NM_153686.8 8217 UTR 3 NP_710153.2
XM_011513821.2 8217 UTR 3 XP_011512123.1
XM_011513822.2 8217 UTR 3 XP_011512124.1
XM_011513823.2 8217 UTR 3 XP_011512125.1
XM_011513825.2 8217 Intron XP_011512127.1
XM_011513826.2 8217 UTR 3 XP_011512128.1
XM_017007961.1 8217 UTR 3 XP_016863450.1
XM_017007962.1 8217 UTR 3 XP_016863451.1
XM_017007963.1 8217 UTR 3 XP_016863452.1
XM_017007964.1 8217 Intron XP_016863453.1
XM_017007965.1 8217 Intron XP_016863454.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 8217 Missense Mutation CCA,CGA P980R NP_071741.2
XM_011513876.2 8217 Missense Mutation CCA,CGA P751R XP_011512178.1
XM_017008543.1 8217 Missense Mutation CCA,CGA P982R XP_016864032.1
XM_017008544.1 8217 Missense Mutation CCA,CGA P753R XP_016864033.1

View Full Product Details