Product Details

SNP ID
rs146988755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69957328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGTCACAGGGTAGATCTGGTG[G/T]GTGGGGTTAAGTAGAAGTTCTTGGT
Phenotype
MIM: 115460
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSN2 PubMed Links

Gene Details

Gene
CSN2
Gene Name
casein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302770.1 637 Silent Mutation NP_001289699.1
NM_001891.3 637 Silent Mutation NP_001882.1
XM_017007760.1 637 Silent Mutation XP_016863249.1

View Full Product Details