Product Details

SNP ID
rs147121215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41990698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGGCCCACAGATGTAGCTGGT[C/T]CTCCCTGTGCCGGCTCCGTCTGCGA
Phenotype
MIM: 604604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF4L1 PubMed Links

Gene Details

Gene
DCAF4L1
Gene Name
DDB1 and CUL4 associated factor 4 like 1
There are no transcripts associated with this gene.

Gene
SLC30A9
Gene Name
solute carrier family 30 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006345.3 193 Missense Mutation TCC,TTC S16F NP_006336.3
XM_017007654.1 193 Missense Mutation TCC,TTC S16F XP_016863143.1

View Full Product Details