Product Details

SNP ID
rs147383260
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:101031649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATCACAAAGAGCTCTGCTGGA[C/T]GGCACAGGTCTAAGGTCTAGAAAGC
Phenotype
MIM: 114105
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PPP3CA PubMed Links

Gene Details

Gene
PPP3CA
Gene Name
protein phosphatase 3 catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000944.4 Intron NP_000935.1
NM_001130691.1 Intron NP_001124163.1
NM_001130692.1 Intron NP_001124164.1
XM_017008365.1 Intron XP_016863854.1

View Full Product Details