Product Details

SNP ID
rs147444946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188139747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTGGAGGACCTTGGAGGGCCC[A/G]CATTTCCAGTCAAACGAGCGTCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIML1 PubMed Links

Gene Details

Gene
TRIML1
Gene Name
tripartite motif family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178556.3 304 Silent Mutation CCA,CCG P63P NP_848651.2
XM_006714207.3 304 Silent Mutation CCA,CCG P63P XP_006714270.1
XM_011531919.1 304 UTR 5 XP_011530221.1

View Full Product Details