Product Details

SNP ID
rs147699912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82356863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCACTGTTGCTGTTGCTGATA[C/T]TGTTCCTTCGACATGGCTACTTTTA
Phenotype
MIM: 601324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPD PubMed Links

Gene Details

Gene
HNRNPD
Gene Name
heterogeneous nuclear ribonucleoprotein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003810.1 1104 Silent Mutation CAA,CAG Q243Q NP_001003810.1
NM_002138.3 1104 Silent Mutation CAA,CAG Q262Q NP_002129.2
NM_031369.2 1104 Silent Mutation CAA,CAG Q243Q NP_112737.1
NM_031370.2 1104 Silent Mutation CAA,CAG Q262Q NP_112738.1

View Full Product Details