Product Details

SNP ID
rs147800526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:79903780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTTTCCTTAACTATGTAACAG[A/T]TGTACCAAGAGTTATTCTATGTTTC
Phenotype
MIM: 608041
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANTXR2 PubMed Links

Gene Details

Gene
ANTXR2
Gene Name
anthrax toxin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145794.1 5579 Intron NP_001139266.1
NM_001286780.1 5579 UTR 3 NP_001273709.1
NM_001286781.1 5579 UTR 3 NP_001273710.1
NM_058172.5 5579 UTR 3 NP_477520.2
XM_011531587.2 5579 Intron XP_011529889.1
XM_017007711.1 5579 Intron XP_016863200.1

View Full Product Details