Product Details

SNP ID
rs148195943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155829848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAAGGAAACTATTTTATTCTA[A/G]GTGACCATTAACACTCCTCTATCCT
Phenotype
MIM: 616693
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASIC5 PubMed Links

Gene Details

Gene
ASIC5
Gene Name
acid sensing ion channel subunit family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017419.2 1573 UTR 3 NP_059115.1
XM_017008291.1 1573 UTR 3 XP_016863780.1

View Full Product Details