Product Details

SNP ID
rs148341393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38773695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATGAAATAAATGCGTGGAATCG[A/G]ACATTTCTCTTGAGTTGTTCTTGGG
Phenotype
MIM: 606270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR10 PubMed Links

Gene Details

Gene
TLR10
Gene Name
toll like receptor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017388.2 2028 Silent Mutation GTC,GTT V632V NP_001017388.1
NM_001195106.1 2028 Silent Mutation GTC,GTT V632V NP_001182035.1
NM_001195107.1 2028 Silent Mutation GTC,GTT V632V NP_001182036.1
NM_001195108.1 2028 Silent Mutation GTC,GTT V618V NP_001182037.1
NM_030956.3 2028 Silent Mutation GTC,GTT V632V NP_112218.2
XM_011513760.2 2028 Silent Mutation GTC,GTT V618V XP_011512062.1
XM_011513761.2 2028 Silent Mutation GTC,GTT V632V XP_011512063.1
XM_011513762.2 2028 Silent Mutation GTC,GTT V632V XP_011512064.1

View Full Product Details