Product Details

SNP ID
rs148434861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70389802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGAATCCCACCTCCTCCTCCCG[C/T]ACCCTATGGTCCAGGGATATTTCCA
Phenotype
MIM: 608936 MIM: 611593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPRPN PubMed Links

Gene Details

Gene
OPRPN
Gene Name
opiorphin prepropeptide
There are no transcripts associated with this gene.

Gene
SMR3B
Gene Name
submaxillary gland androgen regulated protein 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006685.3 343 Missense Mutation GCA,GTA A65V NP_006676.1

View Full Product Details