Product Details

SNP ID
rs149031702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98997338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTGTTAGTATAATTTGGAAAA[A/G]CTTGCACGTGGAGGAGTGTTGCTTG
Phenotype
MIM: 610151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METAP1 PubMed Links

Gene Details

Gene
METAP1
Gene Name
methionyl aminopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015143.2 Intron NP_055958.2
XM_011531777.2 Intron XP_011530079.1
XM_011531778.1 Intron XP_011530080.1
XM_011531779.1 Intron XP_011530081.1
XM_017007914.1 Intron XP_016863403.1
XM_017007915.1 Intron XP_016863404.1
XM_017007916.1 Intron XP_016863405.1
XM_017007917.1 Intron XP_016863406.1
Gene
MIR3684
Gene Name
microRNA 3684
There are no transcripts associated with this gene.

View Full Product Details