Product Details

SNP ID
rs149108321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6576695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTTTTCCGGCTGTGGTGGGAT[G/T]GCGTCGCCTCGGACCAGCAGAAATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAN2B2 PubMed Links

Gene Details

Gene
MAN2B2
Gene Name
mannosidase alpha class 2B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292038.1 293 Missense Mutation GGC,TGC G86C NP_001278967.1
NM_015274.2 293 Missense Mutation GGC,TGC G86C NP_056089.1

View Full Product Details