Product Details

SNP ID
rs149526106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143186129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTTTTTGCAAGCATCTCTTCC[A/T]CAGGTAAGGGTCATTATCTTTCAGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105377623 PubMed Links

Gene Details

Gene
LOC105377623
Gene Name
uncharacterized LOC105377623
There are no transcripts associated with this gene.

Gene
USP38
Gene Name
ubiquitin specific peptidase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290325.1 1213 Missense Mutation ACA,TCA T227S NP_001277254.1
NM_001290326.1 1213 UTR 5 NP_001277255.1
NM_032557.6 1213 Missense Mutation ACA,TCA T227S NP_115946.2
XM_011532360.2 1213 Missense Mutation ACA,TCA T227S XP_011530662.1

View Full Product Details