Product Details

SNP ID
rs149649513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139454361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGACCTACCGCTTCTGCGCT[C/G]GCCGCTTCCCCGACCGCACCGAGGC
Phenotype
MIM: 605950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB33B PubMed Links

Gene Details

Gene
RAB33B
Gene Name
RAB33B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031296.2 651 Missense Mutation CGC,GGC R56G NP_112586.1
XM_011532299.1 651 Missense Mutation CGC,GGC R104G XP_011530601.1

View Full Product Details