Product Details

SNP ID
rs149805235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70158411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATTTACCACCTCCTCTTTATTA[A/T]CGCCCAGTGAATACAGTCCCCAGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRR27 PubMed Links

Gene Details

Gene
PRR27
Gene Name
proline rich 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_214711.3 348 Nonsense Mutation TAA,TAT *53Y NP_999876.2

View Full Product Details