Product Details

SNP ID
rs150270491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184388950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTTGATGGTGACCTGGAGGTC[C/T]GGTTTGTTGGAAGTGACGAAGGACG
Phenotype
MIM: 147576
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF2 PubMed Links

Gene Details

Gene
IRF2
Gene Name
interferon regulatory factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002199.3 1058 Silent Mutation CCA,CCG P286P NP_002190.2
Gene
LOC102723766
Gene Name
uncharacterized LOC102723766
There are no transcripts associated with this gene.

View Full Product Details