Product Details

SNP ID
rs150281141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99905656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCAACTGGCTTAATAATGACAC[A/G]AGGATCAATACAATTATGGGCATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 1322 Silent Mutation CTC,CTT L261L NP_001026893.1
NM_001278310.1 1322 Silent Mutation CTC,CTT L194L NP_001265239.1
NM_001278311.1 1322 Intron NP_001265240.1
XM_011532262.2 1322 Silent Mutation CTC,CTT L176L XP_011530564.1
XM_011532263.2 1322 Silent Mutation CTC,CTT L176L XP_011530565.1
XM_011532265.2 1322 Silent Mutation CTC,CTT L173L XP_011530567.1
XM_017008628.1 1322 Silent Mutation CTC,CTT L173L XP_016864117.1

View Full Product Details