Product Details

SNP ID
rs150426182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153714192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCAAATGGCAGAAGGGGCTCT[A/G]TTCCAACATTTGAAAAATAAATCAT
Phenotype
MIM: 603028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF175 PubMed Links

Gene Details

Gene
RNF175
Gene Name
ring finger protein 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173662.2 Intron NP_775933.1
XM_005262938.3 Intron XP_005262995.1
XM_005262939.3 Intron XP_005262996.1
XM_005262940.4 Intron XP_005262997.1
XM_011531879.2 Intron XP_011530181.1
XM_011531881.2 Intron XP_011530183.1
XM_011531882.2 Intron XP_011530184.1
XM_011531883.2 Intron XP_011530185.1
XM_017008047.1 Intron XP_016863536.1
Gene
TLR2
Gene Name
toll like receptor 2
There are no transcripts associated with this gene.

View Full Product Details