Product Details

SNP ID
rs150641121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70631705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAATGAAGATTCTCCTGGTCTT[C/T]CTAGGGCTTCTTGGTAATTCTGTTG
Phenotype
MIM: 606585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENAM PubMed Links

Gene Details

Gene
ENAM
Gene Name
enamelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031889.2 2120 Silent Mutation TTC,TTT F30F NP_114095.2
XM_006714056.3 2120 Silent Mutation TTC,TTT F30F XP_006714119.1

View Full Product Details