Product Details

SNP ID
rs150722590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67914620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCATAAATTCCTTCCATATATC[C/T]GGCACAGAACATTCCAGGTTTTATA
Phenotype
MIM: 611704
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11A PubMed Links

Gene Details

Gene
TMPRSS11A
Gene Name
transmembrane protease, serine 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114387.1 1184 Missense Mutation AGA,GGA R355G NP_001107859.1
NM_182606.3 1184 Missense Mutation AGA,GGA R358G NP_872412.3

View Full Product Details